Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme D Antibody (RM0246-8G14) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2124090.025MG
Description
Granzyme D Monoclonal specifically detects Granzyme D in Mouse samples. It is validated for Western Blot.Specifications
Granzyme D | |
Monoclonal | |
LYOPH | |
Western Blot 1:100-1:2000 | |
P11033 | |
Gzmd | |
Recombinant protein of Granzyme D. | |
0.025 mg | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2 |
Western Blot | |
RM0246-8G14 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
CCP5, Ctla5, Ctla-5, granzyme D | |
Rat | |
Protein G purified | |
RUO | |
14941 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction