Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granzyme D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567564 Supplier Invitrogen™ Supplier No. PA567564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Granzyme D Polyclonal Antibody for Western Blot

Granzyme D is thought to be necessary for target cell lysis in cell-mediated immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme D
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Gzmd
Gene Accession No. P11033
Gene Alias CCP5; Ctla5; Ctla-5; Granzyme D; Gzmd
Gene Symbols Gzmd
Host Species Rabbit
Immunogen Synthetic peptide derived from mouse Granzyme B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14941
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.