Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme G Antibody (RM0247-12f21) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Granzyme G |
---|---|
Clone | RM0247-12f21 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
Granzyme G Monoclonal specifically detects Granzyme G in Mouse samples. It is validated for Western Blot.Specifications
Granzyme G | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P13366 | |
14944 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0247-12f21 | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Ctla7, Ctla-7 | |
GZMG | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title