Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme G Antibody (RM0247-12f21), mFluor Violet 450 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212410MFV450
Description
Granzyme G Monoclonal antibody specifically detects Granzyme G in Mouse samples. It is validated for Western BlotSpecifications
Granzyme G | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
14944 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0247-12f21 | |
Western Blot | |
Ctla7, Ctla-7 | |
Mouse recombinant Granzyme G | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction