Learn More
Granzyme K Monoclonal Antibody (G3H69), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$383.00
Specifications
| Antigen | Granzyme K |
|---|---|
| Clone | G3H69 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-889-742
|
Invitrogen™
12889742 |
100 Tests |
Each of 1 for $383.00
|
|
|||||
Description
Description: This G3H69 monoclonal antibody reacts with human Granzyme K. Granzyme K is one of five granzyme serine proteases that have been indentified in humans. These proteins are expressed in the granules of NK cells and cytotoxic T cells, and are critical for the induction of target cell apoptosis through the cleavage of intracellular substrates. Granzyme A and Granzyme K are both tryptases and appear to show overlapping, though not identical, substrate specificity. This functional similarity is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTLs. Applications Reported: This G3H69 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G3H69 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.Specifications
| Granzyme K | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| 3003 | |
| IgG2a κ | |
| 4°C, store in dark, DO NOT FREEZE! | |
| GZMK |
| G3H69 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P49863 | |
| GZMK | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.