Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme K Mouse anti-Human, Clone: G3H69, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567938
This item is not returnable.
View return policy
Description
Granzyme K Monoclonal antibody specifically detects Granzyme K in Human samples. It is validated for Intracellular staining (Flow Cytometry)
Specifications
Granzyme K | |
Monoclonal | |
0.5 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
0.1 mg | |
Primary | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
IgG2a κ |
Flow Cytometry | |
G3H69 | |
Unconjugated | |
GZMK; GRAK; TRYP2; fragmentin-3; granzyme 3; tryptase II | |
Human Granzyme K Recombinant Protein | |
RUO | |
Human | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction