Learn More
Granzyme K Mouse anti-Human, Unconjugated, Clone: 1C3A4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Granzyme K |
---|---|
Clone | 1C3A4 |
Concentration | 500 μg/mL |
Applications | Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9403
![]() ![]() |
Proteintech Group Inc
672721IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The Granzyme K antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Granzyme K. This antibody recognizes human antigen. The Granzyme K antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Granzyme K | |
500 μg/mL | |
Monoclonal | |
Mouse | |
3003 | |
Primary |
1C3A4 | |
Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.