Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granzyme K Polyclonal Antibody

Catalog No. PIPA587719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587719 Supplier Invitrogen™ Supplier No. PA587719

Rabbit Polyclonal Antibody

Immunogen sequence: IIGGKEVSPH SRPFMASIQY GGHHVCGGVL IDPQWVLTAA HCQYRFTKGQ SPTVVLGAHS LSKNEASKQT LEIKKFIPFS RVTSDPQSND IMLVKLQTAA KLNKHVKMLH IRSKTSLRSG TKCKVTGWGA TDPDSLRPSD TLREVTVTVL SRKLCNSQSY YNGDPFITKD MVCAGDAKGQ KDSCKGDSGG PLICKGVFHA IVSGGHECGV ATKPGIYTLL TKKYQTWIKS NLVPPHTN; Positive Samples: Mouse liver, Rat liver; Cellular Location: Cytoplasmic granule.

Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme K
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GZMK
Gene Accession No. O35205, P49864
Gene Alias Fragmentin-3; granzyme 3; granzyme K; granzyme K (granzyme 3; tryptase II); granzyme K (serine protease, granzyme 3; tryptase II); granzyme-3; GrK; GZMK; NK-Tryp-2; NK-tryptase-2; RNK-Tryp-2; TRYP2; tryptase II
Gene Symbols GZMK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-264 of human GZMK (NP_0020951).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14945, 29165
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.