Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granzyme K Rabbit anti-Human, Alexa Fluor 750, Clone: 2471A, R&D Systems™

Catalog No. FAB10216S10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10216S10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10216S10 Supplier R&D Systems Supplier No. FAB10216S100UG
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Granzyme K Monoclonal antibody specifically detects Granzyme K in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Granzyme K
Applications Flow Cytometry
Classification Monoclonal
Clone 2471A
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. P49863
Host Species Rabbit
Immunogen Synthetic peptide containing human Granzyme K, Accession # P49863
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.