Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme M Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Granzyme M |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Granzyme M Polyclonal specifically detects Granzyme M in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Granzyme M | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
EC 3.4.21, EC 3.4.21.-, granzyme M, granzyme M (lymphocyte met-ase 1), Hu-Met-1, LMET1, lymphocyte met-ase 1, Met-1 serine protease, MET1met-ase, Met-ase, Natural killer cell granular protease | |
GZMM | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Adaptive Immunity, Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
3004 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: PSMVCLAADSKDQAPCKGDSGGPLVCGKGRVLAGVLSFSSRVCTDIFKPPVATAVAPYVSWIRKVTGR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title