Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granzyme M Monoclonal Antibody (4B2G4), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 53977442
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-977-442 100 Tests
53-977-441 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-977-442 Supplier Invitrogen™ Supplier No. 53977442
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme M
Applications Flow Cytometry
Classification Monoclonal
Clone 4B2G4
Concentration 5 μL/Test
Conjugate Alexa Fluor 488
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene GZMM
Gene Accession No. P51124
Gene Alias granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1
Gene Symbols GZMM
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3004
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Product Line eBioscience
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.