Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Granzyme M Monoclonal Antibody (4B2G4), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$202.00 - $480.00
Specifications
Antigen | Granzyme M |
---|---|
Clone | 4B2G4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-977-441
![]() |
Invitrogen™
12977441 |
25 Tests |
Each for $202.00
|
![]() |
|||||
12-977-442
![]() |
Invitrogen™
12977442 |
100 Tests |
Each for $480.00
|
![]() |
|||||
Description
Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.Specifications
Granzyme M | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1 | |
GZMM | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
GZMM |
4B2G4 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
P51124 | |
3004 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title