Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Granzyme M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100744 Supplier Invitrogen™ Supplier No. PA5100744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GZMM.

Cleaves peptide substrates after methionine, leucine, and norleucine. Physiological substrates include EZR, alpha- tubulins and the apoptosis inhibitor BIRC5/Survivin. Promotes caspase activation and subsequent apoptosis of target cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Granzyme M
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GZMM
Gene Accession No. P51124
Gene Alias granzyme M; granzyme M (lymphocyte met-ase 1); GZMM; Hu-Met-1; Lmet1; lymphoctye Met-ase 1; lymphocyte met-ase 1; MET1; Met-1 serine protease; Met-ase; MMET-1; natural killer cell granular protease; RNK-Met-1
Gene Symbols GZMM
Host Species Rabbit
Immunogen A synthesized peptide derived from human GZMM(Accession P51124), corresponding to amino acid residues G157-S207.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16904, 3004
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.