Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566857 Supplier Thermo Scientific Supplier No. PA566857
Add to Cart
Add to Cart

GRAP Polyclonal Antibody

GRAP Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRAP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRAP
Gene Accession No. Q13588
Gene Alias 8430435N19Rik; BB233594; GRAP; GRB2 related adaptor protein; GRB2-related adapter protein; GRB2-related adaptor protein; growth factor receptor-bound protein 2-related adaptor protein; MGC64880
Gene Symbols GRAP
Host Species Rabbit
Immunogen Recombinant Human GRAP. Recombinant protein control fragment (Product #RP-105650).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.