Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590434
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590434 Supplier Invitrogen™ Supplier No. PA590434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNELVDFYRT TTIAKKRQIF LRDEEPLLKS PGACFAQAQF DFSAQDPSQL SFRRGDIIEV LERPDPHWWR GRSCGRVGFF PRSYVQPVHL; Positive Samples: U-251MG, A-549.

This gene encodes a member of the GRB2/Sem5/Drk family. This member functions as a cytoplasmic signaling protein which contains an SH2 domain flanked by two SH3 domains. The SH2 domain interacts with ligand-activated receptors for stem cell factor and erythropoietin, and facilitates the formation of a stable complex with the BCR-ABL oncoprotein. This protein also associates with the Ras guanine nucleotide exchange factor SOS1 (son of sevenless homolog 1) through its N-terminal SH3 domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRAP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GRAP
Gene Accession No. Q13588
Gene Alias 8430435N19Rik; BB233594; GRAP; GRB2 related adaptor protein; GRB2-related adapter protein; GRB2-related adaptor protein; growth factor receptor-bound protein 2-related adaptor protein; MGC64880
Gene Symbols GRAP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 128-217 of human GRAP (NP_0066041).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10750
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.