Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRAP2 Antibody (OTI1G2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271457B
Description
GRAP2 Monoclonal specifically detects GRAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
GRAP2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GRAP2 | |
Full length recombinant protein of human GRAP2 (NP_004801) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1G2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Adapter protein GRID, GADSSH3-SH2-SH3 adapter Mona, GRB2L, GRB-2-like protein, GRB2-related adapter protein 2, GRB2-related adaptor protein 2, GRB2-related protein with insert domain, GRBLGGRAP-2, GRBX, grf-40, Grf40, Grf40 adapter protein, GRIDGRPL, Growth factor receptor-binding protein, growth factor receptor-bound protein 2-related adaptor protein 2, Hematopoietic cell-associated adapter protein GrpL, hematopoietic cell-associated adaptor protein GRPL, Mona, P38, Protein GADS, SH3-SH2-SH3 adaptor molecule | |
Mouse | |
Affinity Purified | |
Cancer | |
9402.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction