Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRAP2 Antibody (OTI1G2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271457MFV450
Description
GRAP2 Monoclonal antibody specifically detects GRAP2 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
GRAP2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI1G2 | |
Flow Cytometry | |
Adapter protein GRID, GADSSH3-SH2-SH3 adapter Mona, GRB2L, GRB-2-like protein, GRB2-related adapter protein 2, GRB2-related adaptor protein 2, GRB2-related protein with insert domain, GRBLGGRAP-2, GrbX, Grf40, Grf-40, Grf40 adapter protein, GRIDGRPL, Growth factor receptor-binding protein, growth factor receptor-bound protein 2-related adaptor protein 2, Hematopoietic cell-associated adapter protein GrpL, hematopoietic cell-associated adaptor protein GRPL, Mona, P38, Protein GADS, SH3-SH2-SH3 adaptor molecule | |
Full length human recombinant protein of human GRAP2(NP_004801) produced in HEK293T cell. | |
0.1 mL | |
Cancer | |
9402 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction