Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522381 Supplier Invitrogen™ Supplier No. PA522381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat. Predicted reactivity: Mouse (90%), Rat (92%), Rhesus Monkey (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Grap2
Gene Accession No. O75791
Gene Alias adapter protein GRID; Gads; GADS protein; Grap2; GRAP-2; GRB2L; GRB-2-like protein; GRB2-related adapter protein 2; Grb2-related adaptor downstream of Sch; GRB2-related adaptor protein 2; GRB-2-related monocytic adapter protein; GRB2-related protein with insert domain; GRBLG; GRBX; Grf40; grf-40; Grf40 adapter protein; GRID; Growth factor receptor-binding protein; growth factor receptor-bound protein 2-related adaptor protein 2; GrpL; Hematopoietic cell-associated adapter protein GrpL; hematopoietic cell-associated adaptor protein GRPL; MONA; monocytic adapter; monocytic adaptor; P38; Protein GADS; RP3-370M22.1; SH3-SH2-SH3 adapter Mona; SH3-SH2-SH3 adaptor molecule
Gene Symbols Grap2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GRAP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9402
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.