Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite conductive adhesive, alcohol based, Thermo Scientific Chemicals

Catalog No. aa4246515
Click to view available options
Quantity:
30 g

Catalog No. AA4246515

Supplier: Thermo Scientific Chemicals 04246515

Only null left
Add to Cart

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01866817

Specifications

Density 0.9
Boiling Point 82°C
Flash Point 11°C(52°F)
Ignition Point 270°C (518°F)
Quantity 30 g
Vapor Pressure 48hPa (36mm Hg) at 20°C
Chemical Name or Material alcohol based
Hazard Category H225-H318-H335-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.