Learn More
Thermo Scientific Chemicals Graphite Fusion Crucible Lid
Catalog No. AA42315UA
Change view
Click to view available options
Diameter (Metric) Outer:
2.54cm
3.2cm
Quantity:
15/Pk
15/Pk.
Each
Description:
For stock No. 14747
For stock No. 40794
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Quantity Description
AA42315UA 2.54cm 15/Pk For stock No. 40794
AA42315PC 2.54cm Each For stock No. 40794
AA43337UA 3.2cm 15/Pk. For stock No. 14747
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42315UA Supplier Thermo Scientific Chemicals Supplier No. 042315UA
Add to Cart
Graphite is used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description For stock No. 40794
Material Graphite
Quantity 15/Pk
Diameter (Metric) Outer 2.54cm
Product Type Graphite Fusion Crucible Lid