Learn More
Restek™ Graphite O-Rings
SDP
Catalog No. 06611129 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
06-611-129 10/Pk.
1 options
Catalog No. 06-611-129 Supplier Restek™ Supplier No. 21475
For PerkinElmer Auto SYS XL, or Clarus 580/680 GCs with PSS Injector
  • Fits Liners with 4 mm O.D.
Specifications
Diameter (Metric) Outer Fits 4 mm O.D. Liners
For Use With (Application) PerkinElmer Auto SYS XL, or Clarus 580/680 GCs with PSS Injector
Packaging Quantity 10/Pk.
Material Graphite