Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079in), Thermo Scientific Chemicals

Catalog No. p-7059036
Click to view available options
Quantity:
1 Pc.
5 Pc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite plate
Name Note Highly oriented pyrolytic graphite (HOPG), 10x10x(1.6min)mm (0.394x0.394x0.079 in.)
Quantity 1 Pc.
MDL Number MFCD00144065

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.