Alfa Aesar™ Graphite powder, microcrystalline, -325 mesh, 75-82% C, 18-25% Ash

Manufacturer: Alfa Aesar™ 1013014

Quantity 25g

 View more versions of this product

Catalog No. AA1013014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-42-5
    MDL Number MFCD00144065
    UN Number UN1325
    Chemical Name or Material Graphite powder

    Specifications

    Physical Form Micro-Crystalline Powder
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Mesh Size −325 Mesh
    Color Black
    MDL Number MFCD00144065
    CAS 7782-42-5
    UN Number UN1325
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number MD9659600
    Concentration 75 to 82% C, 18 to 25% Ash
    Odor Odorless
    Chemical Name or Material Graphite powder

    GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit