Learn More
Eisco™ Graphite (Pure Carbon) Specimen
SCI_ED
Catalog No. S24669 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24669 1 Ea. 2 to 3 cm
S26621 10/Pk. 2 to 3 cm
S24670 12/Pk. 2 to 3 cm
S26684 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24669 Supplier Eisco™ Supplier No. ESNG0009
Add to Cart
Add to Cart
Graphite specimen correlates well to Next Generation Science Standards Evidence Statements.
  • Example of metallic luster
  • Representation of specimen that is pure carbon and not as expensive as diamond
  • Made of same material found in pencils
  • For use in classroom identification labs and exercises
Specifications
Product Type Mineral
For Use With (Application) Study example of metallic luster
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Graphite