Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphite rod, 6.15mm (0.242in) dia x 102mm (4in) long, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog No. AA40766KD
Click to view available options
Quantity:
50 Pc.

 

Graphite rod is basically used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy. It is mostly used for evaporating carbon onto specimens being prepared for either x-ray microanalysis (EDS or WDS) or transmission electron microscopy (TEM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graphite rod
Name Note 6.15mm (0.242 in.) dia. x 102mm (4 in.) long
Purity Grade Notes AGKSP grade, ultra 'F' purity
Assay Percent Notes (metals basis)
Quantity 50 Pc.
MDL Number MFCD00144065
Solubility Information Insoluble in water.
Percent Purity 99.9995%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.