Alfa Aesar™ Graphite rod, 6.3mm (0.25 in.) dia.. x 61cm (24 in.) long, 99% (metals basis)

Manufacturer: Alfa Aesar™ 10135KI

Quantity 4 pieces

 View more versions of this product

Catalog No. AA10135KI

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00144065
    Chemical Name or Material Graphite rod, 6.3mm (0.25 in.) dia. x 61cm (24 in.) long

    Specifications

    Physical Form Pieces
    MDL Number MFCD00144065
    TSCA TSCA
    Weight ≈40g/pc
    Quantity 4 pieces
    Chemical Name or Material Graphite rod, 6.3mm (0.25 in.) dia. x 61cm (24 in.) long
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit