Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Graphite Sealing Ring
SDP

Catalog No. 06611131 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Graphite
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-611-131 Graphite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-131 Supplier Restek™ Supplier No. 21899
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For Thermo Scientific™ Trace 8000/8000TOP & Focus SSL Instruments; Fits 8 mm O.D. Liners

Specifications

Type Seal
Color Silver
Diameter (English) Outer Fits 8 mm O.D. Liners
For Use With (Application) Thermo Scientific™ Trace 8000/8000 TOP & Focus SSL Instruments
Material Graphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.