Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ GRAPHPACK 2M Ferrules
SDP

Catalog No. 06611134 Shop All Restek Corporation Products
Change view
Click to view available options
Diameter (Metric):
0.25 mm
0.32 mm
0.53 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
06-611-134 0.25 mm
06-611-135 0.32 mm
06-611-136 0.53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-611-134 Supplier Restek™ Supplier No. 22223
Only null left
Add to Cart
Add to Cart

For Gerstel CIS 3 and CIS 4 PTV Inlets

Specifications

For Use With (Equipment) Gerstel CIS 3 and CIS 4 PTV Inlets
Material Brass
Color Brass
For Use With (Application) Gerstel PTV Inlet
Diameter (Metric) 0.25 mm
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.