Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ GRAPHPACK Standard 3D/2 Ferrules
SDP

Catalog No. 06611153 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
For Use With (Equipment):
0.45/0.53 mm Column IDs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-611-153 0.45/0.53 mm Column IDs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-611-153 Supplier Restek™ Supplier No. 20274
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For 0.45/0.53 mm Column IDs, Ideal for GC Applications

  • Provides a complete system that quickly and reliably makes a leak-tight, low-dead-volume connection
  • The central component is a metal-jacketed graphite ferrule
  • Ideal seal for GC applications

Specifications

Size (English) Fitting For 1/16 in., 1/8 in., and 1/4 in.
Diameter (English) Inside 0.8 mm
For Use With (Equipment) 0.45/0.53 mm Column IDs
Material Metal-jacketed Graphite
For Use With (Application) GC
Type Graphite Ferrule
Color Gold
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.