Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRASP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559242 Supplier Invitrogen™ Supplier No. PA559242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEMVTFVCRV HESSPAQLAG LTPGDTIASV NGLNVEGIRH REIVDIIKAS GNVLRLETLY GTSIRKAELE ARLQYLKQTL YEKWG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

GRASP (General receptor for phosphoinositides 1-associated scaffold protein) plays a role in intracellular trafficking and contributes to the macromolecular organization of group 1 metabotropic glutamate receptors (mGluRs) at synapses, linking receptors to neuronal proteins.This protein contains conserved domains, including a leucine zipper sequence, PDZ domain and a C-terminal PDZ-binding motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRASP
Gene Accession No. Q7Z6J2
Gene Alias 95 kDa postsynaptic density protein discs-large ZO-1 domain-containing protein; general receptor for phosphoinositides 1 associated scaffold protein; general receptor for phosphoinositides 1-associated scaffold protein; grasp; GRASP-1; graspl; GRIP1; GRIP-1; GRIPAP1; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein like; Grp1 (general receptor for phosphoinositides)-associated scaffold protein; GRP1-associated scaffold protein; hypothetical protein LOC558950; KIAA1167; LOW QUALITY PROTEIN: general receptor for phosphoinositides 1-associated scaffold protein; MNCb-4428; protein TAMALIN; PSD-95 PDZ domain-containing protein; Tamalin; trafficking regulator and scaffold protein tamalin; zgc:136876
Gene Symbols GRASP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRASP1. Recombinant protein control fragment (Product #RP-97839).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.