Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRASP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569952 Supplier Invitrogen™ Supplier No. PA569952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%.

GRASP (General receptor for phosphoinositides 1-associated scaffold protein) plays a role in intracellular trafficking and contributes to the macromolecular organization of group 1 metabotropic glutamate receptors (mGluRs) at synapses, linking receptors to neuronal proteins.This protein contains conserved domains, including a leucine zipper sequence, PDZ domain and a C-terminal PDZ-binding motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GRASP
Gene Accession No. Q9JJA9
Gene Alias 95 kDa postsynaptic density protein discs-large ZO-1 domain-containing protein; general receptor for phosphoinositides 1 associated scaffold protein; general receptor for phosphoinositides 1-associated scaffold protein; grasp; GRASP-1; graspl; GRIP1; GRIP-1; GRIPAP1; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein; GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein like; Grp1 (general receptor for phosphoinositides)-associated scaffold protein; GRP1-associated scaffold protein; hypothetical protein LOC558950; KIAA1167; LOW QUALITY PROTEIN: general receptor for phosphoinositides 1-associated scaffold protein; MNCb-4428; protein TAMALIN; PSD-95 PDZ domain-containing protein; Tamalin; trafficking regulator and scaffold protein tamalin; zgc:136876
Gene Symbols GRASP
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human GRASP (aa 85-134).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56149
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.