Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRASP1 Antibody (OTI1E3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272064AF594
Description
GRASP1 Monoclonal specifically detects GRASP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GRASP1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
GRIPAP1 | |
Full length recombinant protein of human GRASP1 (NP_064522) produced in HEK293T cells. | |
Neurotransmission | |
56850 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1E3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp434P0630, GRASP1, GRASP-1MGC126595, GRIP1 associated protein 1, GRIP1-associated protein 1, KIAA1167MGC126593, MPMGp800B12492Q3 | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction