Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRASP1 Antibody, R&D Systems™

Catalog No. PPS052
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS052 Supplier R&D Systems Supplier No. PPS052
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prepared from rabbit serum by affinity purification using a column to which the C‑terminal peptide was coupled.

GRASP1 Polyclonal specifically detects GRASP1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot.

Specifications

Antigen GRASP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution
Gene Alias DKFZp434P0630, GRASP1, GRASP-1MGC126595, GRIP1 associated protein 1, GRIP1-associated protein 1, GRIPAP1, KIAA1167MGC126593, MPMGp800B12492Q3
Gene Symbols GRIPAP1
Host Species Rabbit
Immunogen Peptide corresponding to amino acid residues from the C-terminal region of rat GRASP, conjugated to keyhole limpet hemocyanin (KLH)
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56850
Test Specificity This antibody is specific for the 95 kDa GRASP in Western blots of rat cerebellar lysates.
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage For long-term storage, ≤ −20°C is recommended. Product is stable at ≤ −20°C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.