Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRASP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534893
Description
Recommended positive controls: NT2D1, PC-3, SK-N-SH, mouse brain. Predicted reactivity: Mouse (87%), Rat (87%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PDZ domain-containing proteins, such as PSD-95 and GRIP are thought to play key roles in glutamate receptor plasticity. GRIP-associated proteins (GRASPs) that bind to distinct PDZ domains within GRIP also play key roles in regulation of glutamate receptor function. GRASP-1 is a neuronal rasGEF associated with GRIP and AMPA receptors in vivo (Scannevin and Huganir, 2000). Recent work suggests that GRASP-1 may regulate neuronal ras signaling and contribute to the regulation of AMPA receptor distribution by NMDA receptor activity (Ye et al., 2000).
Specifications
| GRASP1 | |
| Polyclonal | |
| Unconjugated | |
| GRIPAP1 | |
| 30kDa C-terminus form; AI854681; DXImx47e; Grasp1; GRASP-1; GRASP-1 C-terminal chain; GRIP1; GRIP-1; GRIP1 associated protein 1; GRIP1-associated protein 1; Gripap1; GRIP-associated protein 1; HCMV-interacting protein; Kiaa1167; mKIAA1167; MPMGp800B12492Q3; Sfc10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54645, 56850 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q4V328, Q8VD04 | |
| GRIPAP1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human GRASP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction