Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRASP55 Antibody (CL2522) - Azide and BSA Free, Novus Biologicals Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP344470
Description
GRASP55 Monoclonal antibody specifically detects GRASP55 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
GRASP55 | |
Monoclonal | |
LYOPH | |
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
Golgi phosphoprotein 6, golgi reassembly stacking protein 2, 55 kDa, golgi reassembly stacking protein 2, 55kDa, Golgi reassembly-stacking protein of 55 kDa, GOLPH6DKFZp434D156, GRASP55FLJ13139, GRS2Golgi reassembly-stacking protein 2, p59 | |
This antibody was generated using a recombinant protein sequence of Q9H8Y8, with the exact immunogen sequence remaining proprietary. | |
100 μg | |
Golgi Apparatus Markers | |
26003 | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
CL2522 | |
Unconjugated | |
Lyophilized from a 0.2 um filtered solution in PBS with Trehalose | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -24°C long term. Avoid freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction