Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRASP55 Monoclonal Antibody (CL2522)
GREENER_CHOICE

Catalog No. PIMA524640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524640 Supplier Invitrogen™ Supplier No. MA524640
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: PTRPFEEGKK ISLPGQMAGT PITPLKDGFT EVQLSSVNPP SLSPPGTTGI EQSLTGLSIS STPPAVSSVL STGV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 88%.

GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP55
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2522
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GORASP2
Gene Accession No. Q9H8Y8
Gene Alias 0610011A07Rik; 2410043M02Rik; 5730520M13Rik; 9430094F20Rik; AW552058; golgi phosphoprotein 6; golgi reassembly stacking protein 2; golgi reassembly stacking protein 2, 55kDa; Golgi reassembly-stacking protein 2; Golgi reassembly-stacking protein of 55 kDa; GOLPH2; GOLPH6; Gorasp2; GRASP55; GRS2; LOW QUALITY PROTEIN: Golgi reassembly-stacking protein 2; p59
Gene Symbols GORASP2
Host Species Mouse
Immunogen Recombinant protein fragment of Human GORASP2 (Product #RP-96030)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26003
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.