Learn More
Invitrogen™ GRASP55 Monoclonal Antibody (CL2610)

Description
Immunogen sequence: SGELLSSLPP TSNAPSDPAT TTAKADAASS LTVDVTPPTA KAPTTVEDRV GDSTPVSEKP VSAAVDANAS ESP Binds to an epitope located within the peptide sequence DAASSLTVDVTPPTA as determined by overlapping synthetic peptides. Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 68%.
Specifications
Specifications
| Antigen | GRASP55 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL2610 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GORASP2 |
| Gene Accession No. | Q9H8Y8 |
| Gene Alias | 0610011A07Rik; 2410043M02Rik; 5730520M13Rik; 9430094F20Rik; AW552058; golgi phosphoprotein 6; golgi reassembly stacking protein 2; golgi reassembly stacking protein 2, 55kDa; Golgi reassembly-stacking protein 2; Golgi reassembly-stacking protein of 55 kDa; GOLPH2; GOLPH6; Gorasp2; GRASP55; GRS2; LOW QUALITY PROTEIN: Golgi reassembly-stacking protein 2; p59 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.