Learn More
GRASP55 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304531AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is involved in establishing the stacked structure of the Golgi apparatus.Specifications
GRASP55 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9H8Y8 | |
GORASP2 | |
Between 400 and 450 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GORASP2 | |
Golgi reassembly-stacking protein 2, GRS2, Golgi reassembly-stacking protein of 55 kDa | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26003 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.