Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRASP65/GORASP1 Mouse anti-Human, Mouse, Rat, Clone: 1B2C10, Proteintech
SDP

Catalog No. 501737069 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7069 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7069 Supplier Proteintech Group Inc Supplier No. 666511IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.

Specifications

Antigen GRASP65/GORASP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B2C10
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GORASP1
Gene Accession No. O35254, Q91X51, Q9BQQ3
Gene Alias FLJ23443, Golgi phosphoprotein 5, GOLPH5, GORASP1, GRASP65, P65
Gene Symbols GORASP1
Host Species Mouse
Immunogen GRASP65,GORASP1 Fusion Protein Ag1054
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56082, 64689, 74498
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.