Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRASP65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593091 Supplier Invitrogen™ Supplier No. PA593091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALPLGAPPPD ALPPGPTPED SPSLETGSRQ SDYMEALLQA PGSSMEDPLP GPGSPSHSAP DPDGLPHFME TPLQPPPPVQ RVMDPGFLDV SGISLLDNSN ASVWPSLPSS TELTTTAVST SGPEDICSSS SSHERGGEAT WSGSEFEVSF LDSPGAQAQA DHLPQLTLPD SLTSAASPED GLSAELLEAQ AEEEPASTEG LDTGTEAEGL DSQAQISTTE; Positive Samples: BT474, U-937; Cellular Location: Cytoplasmic side, Golgi apparatus, Peripheral membrane protein, cis-Golgi network membrane.

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRASP65
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GORASP1
Gene Accession No. Q9BQQ3
Gene Alias 5430411C10Rik; Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; Gorasp1; GRASP65; P65
Gene Symbols GORASP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 221-440 of human GRASP65 (NP_1141051).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64689
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.