Learn More
GRASP65 Polyclonal Antibody, Invitrogen™
Description
PA3-910 detects Grasp65 from human, mouse, and rat samples. PA3-910 has been successfully used in Western blot, IHC (paraffin), immunofluorescence, immunocytochemistry and immunoprecipitation procedures. The PA3-910 immunogen is a GST fustion protein containing the C-terminal residues (202-447) of rat Grasp65.
Specifications
Specifications
| Antigen | GRASP65 |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 0.05% sodium azide |
| Gene | GORASP1 |
| Gene Accession No. | O35254, Q91X51, Q9BQQ3 |
| Gene Alias | 5430411C10Rik; Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; Gorasp1; GRASP65; P65 |
| Gene Symbols | GORASP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.