Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRB14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557131 Supplier Invitrogen™ Supplier No. PA557131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PELCCSPFTS VLSADLFPKA NSRKKQVIKV YSEDETSRAL DVPSDITARD VCQLLILKNH YIDDHSWTLF EHLPHIGVER TIEDHELVIE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 81%.

The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. This protein likely has an inhibitory effect on receptor tyrosine kinase signaling and, in particular, on insulin receptor signaling. This gene may play a role in signaling pathways that regulate growth and metabolism. Transcript variants have been reported for this gene, but their full-length natures have not been determined to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB14
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRB14
Gene Accession No. Q14449
Gene Alias AI505286; GRB14; GRB14 adapter protein; growth factor receptor bound protein 14; growth factor receptor-bound protein 14
Gene Symbols GRB14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRB14. Recombinant protein control fragment (Product #RP-102645).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.