Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRB2 Antibody (M331), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323354
Description
GRB2 Monoclonal antibody specifically detects GRB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
GRB2 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
M331 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:50 | |
abundant SRC homology, Adapter protein GRB2, ASH, EGFRBP-GRB2, epidermal growth factor receptor-binding protein GRB2, Grb3-3, growth factor receptor-bound protein 2, growth factor receptor-bound protein 3, HT027, MST084, MSTP084, NCKAP2, Protein Ash, SH2/SH3 adapter GRB2 | |
Clone (M331) was generated from a full length recombinant rat Grb2 protein. This sequence has high homology to human and mouse Grb2. [UniProt# P62994] | |
0.1 mL | |
Angiogenesis, Breast Cancer, Cancer, Extracellular Matrix, Signal Transduction | |
2885 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction