Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRB7 Monoclonal Antibody (OTI15E7)
GREENER_CHOICE

Catalog No. PIMA527118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527118 Supplier Invitrogen™ Supplier No. MA527118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRB7 Monoclonal Antibody for Western Blot

The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with epidermal growth factor receptor (EGFR) and ephrin receptors. The protein plays a role in the integrin signaling pathway and cell migration by binding with focal adhesion kinase (FAK). Alternative splicing results in multiple transcript variants encoding different isoforms, although the full-length natures of only two of the variants have been determined to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB7
Applications Western Blot
Classification Monoclonal
Clone OTI15E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GRB7
Gene Accession No. Q14451
Gene Alias B47; Epidermal growth factor receptor GRB-7; Grb7; GRB7 adapter protein; growth factor receptor binding protein GRB7; growth factor receptor bound protein 7; growth factor receptor-bound protein 7; Growth factor receptor-bound protein 7 (GRB7 adapter protein) (Epidermal growth factor receptor GRB-7); mKIAA4028; OTTHUMP00000164352
Gene Symbols GRB7
Host Species Mouse
Immunogen Full length human recombinant protein of GRB7 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2886
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.