Learn More
Invitrogen™ GRB7 Polyclonal Antibody

Description
Immunogen sequence: MELDLSPPHL SSSPEDLCPA PGTPPGTPRP PDTPLPEEVK RSQPLLIPTT GRKLREEERR ATSLPSIPNP FPELCSPPSQ SPILGGPSSA RGLLPRDASR PHVVKVYSED GACRSVEVAA GATARHVCEM LVQRAHALSD; Positive Samples: SKOV3, 22Rv1, HeLa; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic granule, Cytoplasmic side, Peripheral membrane protein, focal adhesion.
Specifications
Specifications
| Antigen | GRB7 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.42 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | GRB7 |
| Gene Accession No. | Q14451 |
| Gene Alias | B47; Epidermal growth factor receptor GRB-7; Grb7; GRB7 adapter protein; growth factor receptor binding protein GRB7; growth factor receptor bound protein 7; growth factor receptor-bound protein 7; Growth factor receptor-bound protein 7 (GRB7 adapter protein) (Epidermal growth factor receptor GRB-7); mKIAA4028; OTTHUMP00000164352 |
| Gene Symbols | GRB7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.