Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Great White™ Polyurethane Coated Gloves with Dyneema™ Fibers

Catalog No. 19151255 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-151-255 Small
19-151-258 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-151-255 Supplier PIP™ Supplier No. 19D622
This item is not returnable. View return policy

Dyneema™ fibers are 10 to 15 times stronger than steel, and more durable than polyester with a strength that is 40% greater than aramid fiber. Polyurethane coating on the palm and fingers provides puncture and abrasion resistance.

  • White seamless shell, gray polyurethane coated palm and fingers
  • Washable, resistant to chemicals, water and ultraviolet light
  • Seamless construction
  • Gray polyurethane coated grip for superior puncture and abrasion resistance
  • li>Color coded hems for easy size identification
  • Silicone-free
  • Cut Level 2 protection
  • Dyneema is a brand of polyethylene fibers that are 10 to 15 times stronger than steel, more durable than polyester and a strength that is 40% greater than aramid fiber
  • Cuff Style: Continuous knit

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray, White
Exterior Finish No Texture
Hand Specific
Material Dyneema, Nylon, Spandex
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Construction, Electronics Assembly, Glass Cutting, Sheet Metal Handling, and General Parts Assembly
Glove Cuff Style Continuous Knit
Hem Color Red
Series 19-D622
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.