Learn More
Carolina™ Greater Wax Moth Larvae (Galleria mellonella), Living
SDPSCI_ED
Catalog No. s37291
Click to view available options
Quantity:
50 to 75 Larvae
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Includes prepaid coupon to request delivery of the perishable material later according to convenience.
  • Living Material - Galleria mellonella
  • Moth is easily reared, hardy, odorless, and can be handled safely
  • Unit of 50 to 75 larvae shipped in container with food, with instructions
  • Dimensions: 11.9 x 11.9 x 6.09cm (4.7 x 4.7 x 2.4 in.)
  • Includes wax moth larvae
Specifications
Quantity 50 to 75 Larvae
Size 50 to 75 larvae
Includes Wax moth larvae
Sufficient For Class of 30 students
Product Type Greater Wax Moth Larvae (Galleria mellonella), Living
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.