Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GREB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571960
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571960 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571960 Supplier Invitrogen™ Supplier No. PA571960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GREB1 Polyclonal Antibody for Western Blot

This gene is an estrogen-responsive gene that is an early response gene in the estrogen receptor-regulated pathway. It is thought to play an important role in hormone-responsive tissues and cancer. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GREB1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GREB1
Gene Accession No. Q3UHK3, Q4ZG55
Gene Alias 5730583K22Rik; 9130004E13; AF180470; AU023194; gene regulated by estrogen in breast cancer; gene regulated by estrogen in breast cancer protein; gene regulated in breast cancer 1 protein; GREB1; growth regulating estrogen receptor binding 1; growth regulating estrogen receptor binding 1; protein GREB1; growth regulation by estrogen in breast cancer 1; id:ibd5087; id:ibd5087 protein; im:7140582; Kiaa0575; mKIAA0575; protein GREB1; protein GREB1-like; RGD1564091; si:ch211-175f11.3; zgc:158681
Gene Symbols GREB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1417-1446 amino acids from the Central region of human GREB1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268527, 9687
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.