Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Green Chemistry: Detergents and the Environment
GREENER_CHOICESCI_ED

Catalog No. S96401 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S96401 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96401 Supplier Innovating Science™ Supplier No. IS9502
Add to Cart
Add to Cart

Includes

  • 50mL phosphate builder (10% sodium phosphate) solution
  • 50mL nonphosphate builder (10% sodium silicate/5% citric acid) solution
  • 50mL control (deionized) water, plastic cups (45)
  • Microscope slides (1 box)
  • Coverslips (1 box)
  • Teacher's guide
  • Copy-masters

Requires

  • Natural water sample (collected locally)
  • Aged tap water
  • Marking pens
  • Microscopes
  • Plastic wrap

Understand the consequences of cultural nutrient overloading on the aquatic environment.

  • Recognize that there may be environmentally friendly alternatives in consumer chemical goods
  • Demonstrate and monitor the effects of two detergent builders on natural water samples
  • Visually quantify differences between phosphate and nonphosphate detergent builders on aquatic organisms

Order Info

Class size: 15 students.

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Quantify differences between phosphate/non-phosphate detergent builders on aquatic organisms
Includes 50ml phosphate builder (10% sodium phosphate) solution, 50ml nonphosphate builder (10% sodium silicate/5% citric acid) solution, 50ml control (deionized) water, plastic cups 45, microscope slides 1 box, coverslips 1 box, teacher's guide, copy masters
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 15 Lab Groups
Product Type Detergents and the Environment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.