Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Green Red Cone Opsin, Mouse anti-Human, Clone: 7G8, Millipore Sigma™

Catalog No. MABN249250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN249250 50 μg
MABN2492200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN249250 Supplier MilliporeSigma Supplier No. MABN249250UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Green Red Cone Opsin, clone 7G8, Cat. No. MABN2492, is a highly specific mouse monoclonal antibody that targets Medium-wave-sensitive opsin 1 and Long-wave-sensitive opsin 1 and has been tested for use in Immunohistochemistry (Paraffin).

Medium-wave-sensitive opsin 1/Long-wave-sensitive opsin 1 (UniProts: P04001; P04000; also known as Green cone photoreceptor pigment, Green-sensitive opsin, GOP/ Red cone photoreceptor pigment, Red-sensitive opsin, ROP) are encoded by the OPN1MW (also known as GCP)/OPN1LW (also known as RCP) genes in human. The full range of color discrimination in humans is based on the presence and function of three cone photoreceptors. Each cone type possesses a photo-sensitive pigment-protein complex consisting of 11-cis retinal and a unique opsin protein that gives sensitivity in the short (S cone, peak sensitivity about 420 nm), middle (M cone, peak sensitivity about 530 nm with polymorphism), and long (L cone, peak sensitivity about 560 nm with polymorphism) wavelengths of the light spectrum. Opsins are multi-pass membrane proteins that belongs to the G-protein coupled receptor 1 family. They consist of four extracellular, 7 helical, and four cytoplasmic domains. Genes for the three types of cone opsins and the rod photoreceptor rhodopsin gene seem to be homologous with varying amounts of conservation. Strongest conservation is between the middle (green) and long (red) wavelength sensitive pigments on the X chromosome, suggesting a relatively recent duplication/divergence event. The S cone (blue) opsin seems to have a stronger conservation with rhodopsin. Cone photoreceptor distribution in humans is dominated by the M and L cone pigments. Mutations in OPN1MW and OPN1LW genes are known to cause color blindness that is characterized by a dichromasy in which red and green are confused, without loss of luminance or shift or shortening of the spectrum. Some mutations also lead to cone dystrophy leading to progressive degeneration of the cone photoreceptor with some preservation of rod function. (Ref.: Neitz, M., and Neitz, J. (2000). Arch. Ophthalmol. 118(5); 691-700).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Green Red Cone Opsin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7G8
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Medium-wave-sensitive opsin 1;Green cone photoreceptor pigment;Green-sensitive opsin;GOP;Long-wave-sensitive opsin 1;Red-sensitive opsin;ROP)
Gene Symbols OPN1MW;GCP;OPN1LW;RCP
Host Species Mouse
Immunogen Full-length purified green/red human opsin.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000504.1;NA
Test Specificity Clone 7G8 specifically detects human green/red opsin.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.